missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isothiocyanate, TRC

Product Code. 18053940
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18053940 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18053940 Supplier Toronto Research Chemicals Supplier No. TRCM31497025G
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 556-61-6
Molecular Formula C2 H3 N S
Quantity 25 g
InChI Formula InChI=1S/C2H3NS/c1-3-2-4/h1H3
SMILES CN=C=S
IUPAC Name methylimino(sulfanylidene)methane
Molecular Weight (g/mol) 73.12
Formula Weight 72.9986
Chemical Name or Material Methyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.