Learn More
Methyl Isonicotinate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl Isonicotinate |
| CAS | 2459-09-8 |
| Molecular Formula | C7H7NO2 |
| Quantity | 10 g |
| Synonym | 4-Pyridinecarboxylic Acid Methyl Ester, Isonicotinic Acid Methyl Ester, 4-Carbomethoxypyridine, 4-Methoxycarbonylpyridine, Methyl 4-Pyridinecarboxylate, NSC 18257 |
| InChI Formula | InChI=1S/C7H7NO2/c1-10-7(9)6-2-4-8-5-3-6/h2-5H,1H3 |
| SMILES | COC(=O)c1ccncc1 |
| IUPAC Name | methyl pyridine-4-carboxylate |
| Molecular Weight (g/mol) | 137.14 |
| Formula Weight | 137.0477 |