Learn More
Methyl Glyphosate, TRC
Specifications
Specifications
| Chemical Name or Material | Glyphosate-N-methyl |
| CAS | 24569-83-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C4 H10 N O5 P |
| Quantity | 500 mg |
| Synonym | Glyphosate-N-methyl, N-Methyl-N-(phosphonomethyl)glycine, N-(Phosphonomethyl)-N-methylglycine, N-Methylglyphosate, Glycine','N-methyl-N-(phosphonomethyl)- |
| InChI Formula | InChI=1S/C4H10NO5P/c1-5(2-4(6)7)3-11(8,9)10/h2-3H2,1H3,(H,6,7)(H2,8,9,10) |
| SMILES | CN(CC(=O)O)CP(=O)(O)O |
| IUPAC Name | 2-[methyl(phosphonomethyl)amino]acetic acid |
| Molecular Weight (g/mol) | 183.1 |
| Show More |