Learn More
Methyl Formate, Sigma-Aldrich™
Product Code. 15868450
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15868450 2.5 L 2.5 L
15858450 100 mL 100mL
15826270 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15868450 Supplier Sigma Aldrich Supplier No. 8008892500
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-31-3
Specifications
Melting Point -100.4°C
Density 0.968 g/cm3 at 20°C
Boiling Point 31.5°C/1013 hPa
MDL Number MFCD00003291
Quantity 2.5 L
Synonym Methyl formate, Formic acid methyl ester, Methyl formate
Molecular Weight (g/mol) 60.05
Grade Research