Learn More
Methyl-d3 Acetate, CDN
Specifications
Specifications
| Chemical Name or Material | Methyl Acetate-D3 (methoxy-D3) |
| CAS | 24704-57-2 |
| Molecular Formula | C3 D3 H3 O2 |
| Quantity | 1 g |
| Synonym | Methan-d3-ol, acetate (6 CI, 9 CI), Acetic acid, methyl-d3 ester (8 CI), Methyl-d3 acetate, Methyl Acetate-d3, Methyl Acetate-D3 (methoxy-D3), Acetic acid methyl-d3 ester, Acetic acid methyl ester-d3 |
| InChI Formula | InChI=1 S/C3H6O2/c1-3(4)5-2/h1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])OC(=O)C |
| IUPAC Name | trideuteriomethyl acetate |
| Molecular Weight (g/mol) | 77.097 |
| Formula Weight | 77.0556 g/mol |
| Show More |