Learn More
Methyl-α-D-galactopyranoside, 98+%
Product Code. 10658444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10658444 50 g 50g
10041101 2.5 g 2.5g
10096393 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10658444 Supplier Thermo Scientific Acros Supplier No. 337090500
Request Bulk or Custom Format
CAS: 3396-99-4 | C7H14O6 | 194.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3396-99-4
MDL Number MFCD00064085
PubChem CID 76935
ChEBI CHEBI:55507
Specifications
Melting Point 116.0°C to 117.0°C
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Glass Bottle
Molecular Formula C7H14O6
Quantity 50 g
Specific Rotation Condition + 179.00 (20.00°C c=1,water)
Specific Rotation + 179.00
Synonym methyl alpha-d-galactopyranoside, alpha-methyl-d-galactoside, alpha-methylgalactoside, me-alpha-gal, methyl alpha-d-galactoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol, methyl, a-d-galactopyranoside, methyl-galactopyranoside, methyl-alpha-d-galactopyranoside, alpha-methyl d-galactoside
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
InChI Key HOVAGTYPODGVJG-PZRMXXKTSA-N
SMILES COC1C(C(C(C(O1)CO)O)O)O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Weight (g/mol) 194.18
Formula Weight 194.18
Percent Purity 98+%
Chemical Name or Material Methyl-α-D-galactopyranoside