missing translation for 'onlineSavingsMsg'
Learn More

Methyl-α-D-galactopyranoside, 98+%

Product Code. 10096393 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10096393 10 g 10g
10041101 2.5 g 2.5g
10658444 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096393 Supplier Thermo Scientific Acros Supplier No. 337090100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3396-99-4 | C7H14O6 | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-99-4
MDL Number MFCD00064085
PubChem CID 76935
ChEBI CHEBI:55507

Specifications

Assay Percent Range 98+%
Molecular Formula C7H14O6
Quantity 10 g
Specific Rotation Condition + 179.00 (20.00°C c=1,water)
Synonym methyl alpha-d-galactopyranoside, alpha-methyl-d-galactoside, alpha-methylgalactoside, me-alpha-gal, methyl alpha-d-galactoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol, methyl, a-d-galactopyranoside, methyl-galactopyranoside, methyl-alpha-d-galactopyranoside, alpha-methyl d-galactoside
InChI Key HOVAGTYPODGVJG-PZRMXXKTSA-N
SMILES COC1C(C(C(C(O1)CO)O)O)O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular Weight (g/mol) 194.183
Formula Weight 194.18
Chemical Name or Material Methyl-α-D-galactopyranoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.