Learn More
Methyl cyclopentanecarboxylate, 97%, BLD Pharm™
Product Code. 31007351
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31007351 10 g 10g
31005212 5 g 5g
30999762 1 g 1g
31006821 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31007351 Supplier BLD Pharm Supplier No. BD13940310g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 4630-80-2
Molecular Formula C7H12O2
MDL Number MFCD00019290
Quantity 10 g
SMILES O=C(C1CCCC1)OC
Molecular Weight (g/mol) 128.17
Percent Purity 97%
Chemical Name or Material Methyl cyclopentanecarboxylate