missing translation for 'onlineSavingsMsg'
Learn More

Methyl Alpha-D-Galactopyranoside, TRC

Product Code. 30473137
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473137 50 g 50g
30469124 25 g 25g
30479240 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473137 Supplier Toronto Research Chemicals Supplier No. TRCM30550050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3396-99-4
Molecular Formula C7 H14 O6
Molecular Weight (g/mol) 194.18
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3/t3-,4+,5+,6-,7+/m1/s1
Formula Weight 194.079
Chemical Name or Material Methyl-alpha-D-galactopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.