missing translation for 'onlineSavingsMsg'
Learn More

Methyl Acrylate, Sigma-Aldrich™

Product Code. 15846260
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15846260 100 mL 100mL
15838430 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15846260 Supplier Sigma Aldrich Supplier No. 8008410100
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized with hydroquinone monomethyl ether for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-33-3

Specifications

Melting Point −75°C (lit.)
Density 0.956 g/mL at 25°C
Boiling Point 80°C
Molecular Formula CH2=CHCOOCH3
MDL Number MFCD00008627
Quantity 100 mL
Synonym Methyl acrylate, Methyl propenoate, Acrylic acid methyl ester, Methyl acrylate
Molecular Weight (g/mol) 86.09
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.