Learn More
Methyl 6-amino-2-naphthoate, 95%, BLD Pharm™
Product Code. 31007248
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31007248 100 mg 100mg
31001689 5 g 5g
30990512 25 g 25g
30998932 250 mg 250mg
31005547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31007248 Supplier BLD Pharm Supplier No. BD108096100mg
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 5159-59-1
Molecular Formula C12H11NO2
MDL Number MFCD06797040
Quantity 100 mg
SMILES O=C(OC)C1=CC=C2C=C(N)C=CC2=C1
Molecular Weight (g/mol) 201.22
Percent Purity 95%
Chemical Name or Material Methyl 6-amino-2-naphthoate