missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-chloro-5-oxovalerate, 97%

Product Code. 11476207
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476207 250 g 250g
11456207 10 g 10g
11466207 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476207 Supplier Thermo Scientific Alfa Aesar Supplier No. B25141.30
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1501-26-4 | C6H9ClO3 | 164.59 g/mol

Methyl 5-chloro-5-oxovalerate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-chloro-5-oxovalerate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1501-26-4
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000756
InChI Key JCAZSWWHFJVFPP-UHFFFAOYSA-N
Synonym methyl 4-chloroformyl butyrate, methyl 5-chloro-5-oxovalerate, methyl 4-chloroformylbutyrate, methyl5-chloro-5-oxopentanoate, 5-chloro-5-oxopentanoic acid methyl ester, methyl glutaryl chloride, pentanoic acid, 5-chloro-5-oxo-, methyl ester, 4-chlorocarbonyl-butyric acid methyl ester, zlchem 559, pubchem14033
PubChem CID 73916
IUPAC Name methyl 5-chloro-5-oxopentanoate
SMILES COC(=O)CCCC(Cl)=O

Specifications

Boiling Point 108°C to 110°C (15 mmHg)
Linear Formula ClCO(CH2)3CO2CH3
Refractive Index 1.447
Quantity 250 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 164.59
Percent Purity 97%
Chemical Name or Material Methyl 5-chloro-5-oxovalerate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.