Learn More
Methyl 5-chloro-5-oxovalerate, 97%
Product Code. 11456207
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456207 10 g 10g
11466207 50 g 50g
11476207 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11456207 Supplier Thermo Scientific Alfa Aesar Supplier No. B25141.09
 Restricted Item
In Stock
Add to basket
CAS: 1501-26-4 | C6H9ClO3 | 164.59 g/mol
Methyl 5-chloro-5-oxovalerate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 5-chloro-5-oxovalerate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases and moisture.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1501-26-4
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000756
InChI Key JCAZSWWHFJVFPP-UHFFFAOYSA-N
Synonym methyl 4-chloroformyl butyrate, methyl 5-chloro-5-oxovalerate, methyl 4-chloroformylbutyrate, methyl5-chloro-5-oxopentanoate, 5-chloro-5-oxopentanoic acid methyl ester, methyl glutaryl chloride, pentanoic acid, 5-chloro-5-oxo-, methyl ester, 4-chlorocarbonyl-butyric acid methyl ester, zlchem 559, pubchem14033
PubChem CID 73916
IUPAC Name methyl 5-chloro-5-oxopentanoate
SMILES COC(=O)CCCC(Cl)=O
Specifications
Boiling Point 108°C to 110°C (15 mmHg)
Linear Formula ClCO(CH2)3CO2CH3
Refractive Index 1.447
Quantity 10 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 164.59
Percent Purity 97%
Chemical Name or Material Methyl 5-chloro-5-oxovalerate