Learn More
Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 50593-91-4 |
| Molecular Formula | C7H7BrN2O2S |
| MDL Number | MFCD05664428 |
| Quantity | 10 g |
| SMILES | O=C(C1=NC(SC)=NC=C1Br)OC |
| Molecular Weight (g/mol) | 263.11 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate |