Learn More
Methyl 4-amino-3-chlorobenzoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 84228-44-4 |
| Molecular Formula | C8H8ClNO2 |
| MDL Number | MFCD01320686 |
| Quantity | 1 g |
| SMILES | COC(=O)C1=CC(Cl)=C(N)C=C1 |
| Molecular Weight (g/mol) | 185.61 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 4-amino-3-chlorobenzoate |