Learn More
Methyl 4-(Acetylamino)-2-hydroxybenzoate, Mikromol™
Product Code. 17978754
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978754 100 mg 100mg
17987314 25 mg 25mg
30458769 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17978754 Supplier Mikromol Supplier No. MM0073.10
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-(Acetylamino)-2-hydroxybenzoate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 4-(Acetylamino)-2-hydroxybenzoate
CAS 4093-28-1
Analyte or Component Names Methyl 4-(Acetylamino)-2-hydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C10 H11 N O4
Certifications/Compliance ISO 9001
Synonym Methyl 4-(Acetylamino)-2-hydroxybenzoate, Ethopabate Related Compound A
InChI Formula InChI=1S/C10H11NO4/c1-6(12)11-7-3-4-8(9(13)5-7)10(14)15-2/h3-5,13H,1-2H3,(H,11,12)
SMILES COC(=O)c1ccc(NC(=O)C)cc1O
IUPAC Name methyl 4-acetamido-2-hydroxybenzoate
Molecular Weight (g/mol) 209.2
Quantity 100 mg
Formula Weight 209.0688 g/mol