Learn More
Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate, Mikromol™
Description
Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate
Specifications
Specifications
| Chemical Name or Material | Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate |
| CAS | 155405-80-4 |
| Analyte or Component Names | Methyl 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H16 N4 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid Methyl Ester, Benzoic acid, 4-[2-(2-amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]-, methyl ester, Benzoic acid, 4-[2-(2-amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]-, methyl ester (9CI), 1H-Pyrrolo[2,3-d]pyrimidine, benzoic acid deriv. |
| InChI Formula | InChI=1S/C16H16N4O3/c1-23-15(22)10-5-2-9(3-6-10)4-7-11-8-18-13-12(11)14(21)20-16(17)19-13/h2-3,5-6,8H,4,7H2,1H3,(H4,17,18,19,20,21) |
| SMILES | COC(=O)c1ccc(CCc2c[nH]c3NC(=NC(=O)c23)N)cc1 |
| IUPAC Name | methyl 4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoate |
| Show More |