Learn More
Methyl 3-(methylthio)propionate, TRC
Product Code. 30476610
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476610 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30476610 Supplier Toronto Research Chemicals Supplier No. TRCM2386452.5G
 Restricted Item
Estimated Shipment: 26-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 3-(methylthio)propionate
CAS 13532-18-8
Molecular Formula C5 H10 O2 S
Quantity 2.5 g
InChI Formula InChI=1S/C5H10O2S/c1-7-5(6)3-4-8-2/h3-4H2,1-2H3
SMILES COC(=O)CCSC
IUPAC Name methyl 3-methylsulfanylpropanoate
Molecular Weight (g/mol) 134.197
Formula Weight 134.04