missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(methylthio)propionate, 99+%, Thermo Scientific™

Product Code. p-165317 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712243 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10712243 Supplier Thermo Scientific Acros Supplier No. 147000050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13532-18-8
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.19
MDL Number MFCD00008565
InChI Key DMMJVMYCBULSIS-UHFFFAOYSA-N
Synonym methyl 3-methylthio propionate, methyl 3-methylthiopropionate, methyl 3-methylthio propanoate, propanoic acid, 3-methylthio-, methyl ester, methyl beta-methiopropionate, methyl 3-methylsulfanyl propanoate, methyl-3-methylthio propionate, methyl 3-methylthiopropanoate, propionic acid, 3-methylthio-, methyl ester, methyl beta-methylthiopropionate
PubChem CID 61641
IUPAC Name methyl 3-methylsulfanylpropanoate
SMILES COC(=O)CCSC

Specifications

Color Colorless
Density 1.0700g/mL
Boiling Point 74.0°C to 75.0°C (13.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3SCH2CH2CO2CH3
Refractive Index 1.4640 to 1.4660
Quantity 5 g
Specific Gravity 1.07
Solubility Information Solubility in water: insoluble.
Formula Weight 134.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Methyl 3-(methylthio)propionate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.