missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3,5-dinitrobenzoate, 98%

Product Code. 11466126
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466126 250 g 250g
11456126 10 g 10g
11418820 25 g 25g
11428820 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11466126 Supplier Alfa Aesar™ Supplier No. B22973.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2702-58-1
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.144
MDL Number MFCD00017016
InChI Key POGCCFLNFPIIGW-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-dinitro-, methyl ester, 3,5-dinitro-benzoic acid methyl ester, methyl 3,5-dinitrobenzoate, methyl-3,5-dinitrobenzoate, acmc-1cr6q, d0x5an, ksc494q3h, methyl-3,5-dinitro benzoate, 3,5-dinitrobenzoic acid methyl ester
PubChem CID 222067
IUPAC Name methyl 3,5-dinitrobenzoate
SMILES COC(=O)C1=CC(=CC(=C1)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 108°C to 112°C
Quantity 250 g
Beilstein 1252593
Formula Weight 226.15
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dinitrobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.