Learn More
Methyl 3-(1-Tritylimidazol-4-yl) Propionate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 3-(1-Tritylimidazol-4-yl) Propionate |
| CAS | 102676-60-8 |
| Molecular Formula | C26 H24 N2 O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C26H24N2O2/c1-30-25(29)18-17-24-19-28(20-27-24)26(21-11-5-2-6-12-21,22-13-7-3-8-14-22)23-15-9-4-10-16-23/h2-16,19-20H,17-18H2,1H3 |
| SMILES | COC(=O)CCc1cn(cn1)C(c2ccccc2)(c3ccccc3)c4ccccc4 |
| IUPAC Name | methyl 3-(1-tritylimidazol-4-yl)propanoate |
| Molecular Weight (g/mol) | 396.48 |
| Formula Weight | 396.1838 |