Learn More
Methyl 3-(1-Tritylimidazol-4-yl) Propionate, TRC
Product Code. 30480394
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480394 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30480394 Supplier Toronto Research Chemicals Supplier No. TRCM3317751G
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 3-(1-Tritylimidazol-4-yl) Propionate
CAS 102676-60-8
Molecular Formula C26 H24 N2 O2
Quantity 1 g
InChI Formula InChI=1S/C26H24N2O2/c1-30-25(29)18-17-24-19-28(20-27-24)26(21-11-5-2-6-12-21,22-13-7-3-8-14-22)23-15-9-4-10-16-23/h2-16,19-20H,17-18H2,1H3
SMILES COC(=O)CCc1cn(cn1)C(c2ccccc2)(c3ccccc3)c4ccccc4
IUPAC Name methyl 3-(1-tritylimidazol-4-yl)propanoate
Molecular Weight (g/mol) 396.48
Formula Weight 396.1838