missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-isothiocyanatoacetate, 98%

Product Code. 11397506
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397506 5 g 5g
11387506 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397506 Supplier Thermo Scientific Alfa Aesar Supplier No. L11046.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 21055-37-8 | C4H5NO2S | 131.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21055-37-8
Molecular Formula C4H5NO2S
Molecular Weight (g/mol) 131.149
MDL Number MFCD00041119
InChI Key GOWGDPFDGIPFIK-UHFFFAOYSA-N
Synonym methyl isothiocyanatoacetate, acetic acid, isothiocyanato-, methyl ester, acmc-20akih, propargyl benzene sulphonate, methyl-2-isothiocyanatoacetate, methyl isothiocyanatoacetate #, methyl 2-isothiocyanatoethanoate, methyl n-thioxomethylene glycinate, isothiocyanatoacetic acid methyl ester, 2-isothiocyanatoacetic acid methyl ester
PubChem CID 140795
IUPAC Name methyl 2-isothiocyanatoacetate
SMILES COC(=O)CN=C=S

Specifications

Density 1.21
Boiling Point 137°C to 138°C (35 mmHg)
Odor Irritating
Linear Formula SCNCH2CO2CH3
Refractive Index 1.519
Quantity 5 g
UN Number UN2922
Beilstein 2074764
Sensitivity Moisture sensitive
Formula Weight 131.15
Percent Purity 98%
Chemical Name or Material Methyl 2-isothiocyanatoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.