Learn More
Methyl 2-isothiocyanatoacetate, 98%
Product Code. 11387506
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387506 1 g 1g
11397506 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11387506 Supplier Thermo Scientific Alfa Aesar Supplier No. L11046.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
CAS: 21055-37-8 | C4H5NO2S | 131.149 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21055-37-8
Molecular Formula C4H5NO2S
Molecular Weight (g/mol) 131.149
MDL Number MFCD00041119
InChI Key GOWGDPFDGIPFIK-UHFFFAOYSA-N
Synonym methyl isothiocyanatoacetate, acetic acid, isothiocyanato-, methyl ester, acmc-20akih, propargyl benzene sulphonate, methyl-2-isothiocyanatoacetate, methyl isothiocyanatoacetate #, methyl 2-isothiocyanatoethanoate, methyl n-thioxomethylene glycinate, isothiocyanatoacetic acid methyl ester, 2-isothiocyanatoacetic acid methyl ester
PubChem CID 140795
IUPAC Name methyl 2-isothiocyanatoacetate
SMILES COC(=O)CN=C=S
Specifications
Density 1.21
Boiling Point 137°C to 138°C (35 mmHg)
Odor Irritating
Linear Formula SCNCH2CO2CH3
Refractive Index 1.519
Quantity 1 g
UN Number UN2922
Beilstein 2074764
Sensitivity Moisture sensitive
Formula Weight 131.15
Percent Purity 98%
Chemical Name or Material Methyl 2-isothiocyanatoacetate