missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-2-deoxy-b-D-glucopyranoside HCl, TRC

Product Code. 30479015
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479015 2 g 2g
30468059 500 mg 500mg
30468360 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479015 Supplier Toronto Research Chemicals Supplier No. TRCM2083502G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3867-93-4
Molecular Formula C7H15NO5 HCl
Molecular Weight (g/mol) 229.66
Synonym β-D-Glucopyranoside, methyl 2-amino-2-deoxy-, hydrochloride (1:1) (ACI), Glucopyranoside, methyl 2-amino-2-deoxy-, hydrochloride, β-D- (6CI, 7CI), β-D-Glucopyranoside, methyl 2-amino-2-deoxy-, hydrochloride (9CI), Methyl 2-amino-2-deoxy-β-D-glucopyranoside monohydrochloride, Methyl β-D-glucosaminide monohydrochloride, Methyl 2-amino-2-deoxy-β-D-glucopyranoside hydrochloride
IUPAC Name (2R,3S,4R,5R,6R)-5-amino-2-(hydroxymethyl)-6-methoxyoxane-3,4-diol;hydrochloride
SMILES Cl.CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1N

Specifications

Quantity 2 g
InChI Formula InChI=1S/C7H15NO5.ClH/c1-12-7-4(8)6(11)5(10)3(2-9)13-7;/h3-7,9-11H,2,8H2,1H3;1H/t3-,4-,5-,6-,7-;/m1./s1
Formula Weight 229.0717
Chemical Name or Material Methyl 2-amino-2-deoxy-b-D-glucopyranoside HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.