CAS RN 3867-93-4
CAS RN 3867-93-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-amino-2-deoxy-b-D-glucopyranoside HCl, TRC
CAS: 3867-93-4 Molecular Formula: C7H15NO5 HCl Molecular Weight (g/mol): 229.66 Synonym: β-D-Glucopyranoside, methyl 2-amino-2-deoxy-, hydrochloride (1:1) (ACI),Glucopyranoside, methyl 2-amino-2-deoxy-, hydrochloride, β-D- (6CI, 7CI),β-D-Glucopyranoside, methyl 2-amino-2-deoxy-, hydrochloride (9CI),Methyl 2-amino-2-deoxy-β-D-glucopyranoside monohydrochloride,Methyl β-D-glucosaminide monohydrochloride,Methyl 2-amino-2-deoxy-β-D-glucopyranoside hydrochloride IUPAC Name: (2R,3S,4R,5R,6R)-5-amino-2-(hydroxymethyl)-6-methoxyoxane-3,4-diol;hydrochloride SMILES: Cl.CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1N