missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MethoCult™ H4034 Optimum

Product Code. 15598309
Encompass
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15598309 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15598309 Supplier STEMCELL Technologies Supplier No. 04034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methylcellulose-based medium with recombinant cytokines for human cells

  • MethoCult™ H4034 Optimum (MethoCult™ GF H4034) is a methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples
  • MethoCult™ H4034 Optimum has been formulated to support optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage, megakaryocyte progenitor cells (CFU-GEMM)
  • The inclusion of G-CSF in Optimum formulations improves discrimination of myeloid colony subtypes
  • This formulation is compatible with STEMvision™ software for automated colony counting

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.