missing translation for 'onlineSavingsMsg'
Learn More

Methapyrilene Dihydrochloride, TRC

Product Code. 30467170
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467170 500 mg 500mg
30465296 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30467170 Supplier Toronto Research Chemicals Supplier No. TRCM259945500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methapyrilene Dihydrochloride
Molecular Formula C14H19N3S . 2(HCl)
Quantity 500 mg
Synonym N1,N1-Dimethyl-N2-2-pyridinyl-N2-(2-thienylmethyl)-1,2-ethanediamine Dihydrochloride, Thenylene Dihydrochloride, Thenylpyramine Dihydrochloride
InChI Formula InChI=1S/C14H19N3S/c1-16(2)9-10-17(12-13-6-5-11-18-13)14-7-3-4-8-15-14/h3-8,11H,9-10,12H2,1-2H3
SMILES CN(C)CCN(Cc1cccs1)c2ccccn2
Molecular Weight (g/mol) 261.39
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.