Learn More
Methanol, Supelco™
Gradient grade for liquid chromatography LiChrosolv™ Reag. Ph Eur, Reag. ChP
66.04 EUR - 4960.00 EUR
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
15804730
|
Supelco
1060072500 |
121.00 EUR 66.04 EUR / 2.5 L Save 54.96 EUR 45% Off |
In Stock
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15814730
|
Supelco
1060079010 |
444.00 EUR
10 L |
Estimated Shipment: 08-01-2027
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15824730
|
Supelco
1060079030 |
948.00 EUR
30 L |
Estimated Shipment: 08-01-2027
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15807420
|
Supelco
1060079185 |
4960.00 EUR
185 L |
Estimated Shipment: 13-11-2026
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
LiChrosolv™ solvents were specially formulated to meet the demanding needs of analytical HPLC applications. With exceptional UV transmittance, minimal particle count, and controlled acidity and alkalinity, our solvents ensure reproducible separations every time. The low evaporation residue level further enhances their performance, making them the ideal choice for high-quality results. The product line includes both gradient grade and isocratic grade solvents, specifically designed to reduce the gradient effect during separations. This tailored approach allows for optimal consistency and accuracy in your analytical processes, helping you achieve the best possible outcomes in your chromatography work. Now Available with Data Matrix Code This high-quality HPLC-grade solvent now features a unique Data Matrix Code on its label, providing digital access to essential product information such as batch number, product number, and expiry date. Unlike standard barcodes, our DMCs feature a unique serial number for each bottle, enabling you to monitor and trace every bottle individually. With standard barcodes, all bottles from the same batch look identical. Our solution gives you true bottle-level traceability for greater control and compliance in the lab. Know more about our data matrix code-labeled HPLC-grade solvents.
Safety and Handling
- Methanol gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur,Reag. ChP
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
- H370-Causes damage to organs.
Precautionary Statement
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P311-Call a POISON CENTER/doctor/ .
- P330-Rinse mouth.