missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, GC Ultra-Trace Analysis Grade, Scharlab
Description
GC Ultra-Trace Analysis Grade products are extremely purified solvents that are suitable for the analysis of high, medium and low volatile halogenated hydrocarbons and polyaromatic hydrocarbons.
- Suitable for residue analysis of organohalogenated pesticide and dioxins, furans, PCBs and polycyclic aromatic hydrocarbons
- GC-ECD (from dichloromethane to 1,2,4-trichlorobenze): produced no peaks greater than 1ng/mL as tetrachloromethane; no peaks in the vicinity of 2,4,5-trichlorobiphenyl
- GC-FID (from 1-decanol to 1-tetradecanol): produced no peaks over 5ng/mL as 1-tetradecanol; no peaks in the vicinity of pyrene
Specifications
Specifications
| Melting Point | -98°C |
| CAS | 67-56-1 |
| Color | Colorless |
| Density | 0.79g/cm3 |
| Quantity | 2.5 L |
| Flash Point | 11°C |
| Identification | Passes test |
| Ignition Point | 455°C |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?