Learn More
Methanesulfonic acid methyl ester, 99.4%, For GC analysis, MP Biomedicals
Product Code. 11487672 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487672 25 g 25g
11477672 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11487672 Supplier MP Biomedicals Supplier No. 0215538425
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 66-27-3
Molecular Formula C2H6O3S
Molecular Weight (g/mol) 110.127
InChI Key MBABOKRGFJTBAE-UHFFFAOYSA-N
Synonym methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
PubChem CID 4156
ChEBI CHEBI:25255
IUPAC Name methyl methanesulfonate
SMILES COS(=O)(=O)C
Specifications
Color Colorless
Density 1.3g/mL
Quantity 25 g
Formula Weight 110.13
Percent Purity 0.994
Physical Form Liquid
Chemical Name or Material Methanesulfonic acid methyl ester, For GC analysis