missing translation for 'onlineSavingsMsg'
Learn More

γ-Methacryloxypropyltrimethoxysilane, 99.04%, MP Biomedicals™

Product Code. 11407672 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407672 5 mL 5mL
11417672 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11407672 Supplier MP Biomedicals Supplier No. 0215537805
  • ONLINE EXCLUSIVE PRICE
    58.45 EUR
    55.65 EUR / 5mL
    Save 2.80 EUR
    4% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2530-85-0
Molecular Formula C10H20O5Si
Molecular Weight (g/mol) 248.35
MDL Number MFCD00008593
InChI Key XDLMVUHYZWKMMD-UHFFFAOYSA-N
Synonym 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
PubChem CID 17318
IUPAC Name 3-trimethoxysilylpropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC[Si](OC)(OC)OC

Specifications

Color Colorless
Density 1.04g/mL
Quantity 5 mL
Formula Weight 248.35
Percent Purity 0.9904
Physical Form Liquid
Chemical Name or Material γ-Methacryloxypropyl-trimethoxysilane, Adhesive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.