missing translation for 'onlineSavingsMsg'
Learn More

Methacrylic Acid Alkylester (Mixture of C12-C16), Sigma-Aldrich™

Product Code. 15854001
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854001 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15854001 Supplier Sigma-Aldrich Supplier No. 8005890500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

(Stabilized with hydroquinone monomethyl ether for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90551-76-1

Specifications

Melting Point -25°C
Density 0.87 g/cm3 at 20°C
MDL Number MFCD01121974
Quantity 500 mL
Synonym Methacrylic acid alkylester (mixture of C12-C16), Dodecyl 2-methylpropenoate, Lauryl methacrylate, Methacrylic acid dodecyl ester, Methacrylic acid alkylester (mixture of C12-C16)
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.