Learn More
Metconazole-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Metconazole-d6 |
| CAS | 2469554-74-1 |
| Molecular Formula | C17H16D6ClN3O |
| Quantity | 1 mg |
| Synonym | Metconazole d6, Metconazole (D6) |
| InChI Formula | InChI=1S/C17H22ClN3O/c1-16(2)8-7-14(9-13-3-5-15(18)6-4-13)17(16,22)10-21-12-19-11-20-21/h3-6,11-12,14,22H,7-10H2,1-2H3/i1D3,2D3 |
| SMILES | ClC1=CC=C(CC2C(O)(CN3C=NC=N3)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])CC2)C=C1 |
| IUPAC Name | 5-[(4-chlorophenyl)methyl]-1-(1,2,4-triazol-1-ylmethyl)-2,2-bis(trideuteriomethyl)cyclopentanol |
| Molecular Weight (g/mol) | 325.87 |
| Formula Weight | 325.18 |