Learn More
Metanil Yellow, Supelco™
Specifications
Specifications
| CAS | 587-98-4 |
| MDL Number | MFCD00007486 |
| Synonym | Metanil yellow, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, Metanil yellow |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 375.38 |
| Grade | Research |
| Chemical Name or Material | Metanil Yellow |