Learn More
Sampling Systems™ Metal Detectable V Spatulas
Product Code. 17524716 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric) Blade:
150 mm
250 mm
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade unitSize
17524716 250 mm Pack of 4
17514716 150 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17524716 Supplier Sampling Systems™ Supplier No. M1010B250
Only null left
Add to basket
Add to basket
Sampling powder is easy with Metal Detectable V Spatulas. V-Spatulas are ideal for both sampling and dispensing and are suitable for use with powders and granules.
  • Metal Detectable V Spatulas allow for fast powder sampling.
  • They are made from a special tough, chemically resistant grade of PP.
  • The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
  • This ensures the cleanliness and purity of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • Metal Detectable V-Spatulas are used for surface sampling from drums and kegs and they can also be used for taking samples through sacks.
  • Our ergonomic handle means they are comfortable to use.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • 150 mm and 250 mm blade length is a standard size.
Specifications
Color Blue
Length (Metric) Blade 250 mm