Learn More
Picloram in Methanol 100μg/mL, Fisher Chemical™
Product Code. 12946413 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12946413 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12946413 Supplier Fisher Chemical Supplier No. P848520
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule
CAS 2-1-1918
Physical Form Liquid
Molecular Formula C6H3Cl3N2O2
Concentration 100 μg/mL
InChI Key NQQVFXUMIDALNH-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N
IUPAC Name 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
Solvent or Matrix Methanol
Molecular Weight (g/mol) 241.45
PubChem CID 15965
Quantity 1 mL
ChEBI CHEBI:34922
Chemical Name or Material Picloram
Show More Show Less
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.