missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Endosulfan in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 100μg/mL |
CAS | 33213-65-9 |
Packaging | Ampoule(s) |
Quantity | 1 mL |
Molecular Formula | C9H6Cl6O3S |
MDL Number | MFCD00137651 |
InChI Key | RDYMFSUJUZBWLH-VQDHCCOSSA-N |
SMILES | ClC1=C(Cl)[C@@]2(Cl)[C@H]3COS(=O)OC[C@H]3C1(Cl)C2(Cl)Cl |
IUPAC Name | (1S,2R,8S)-1,9,10,11,12,12-hexachloro-4,6-dioxa-5λ⁴-thiatricyclo[7.2.1.0²,⁸]dodec-10-en-5-one |
Molecular Weight (g/mol) | 406.90 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'