missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite Beta-Endosulfan in Acetonitrile 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 100μg/mL |
CAS | 33213-65-9 |
Packaging | Ampoule(s) |
Quantity | 1 mL |
Molecular Formula | C9H6Cl6O3S |
MDL Number | MFCD00137651 |
UN Number | 1648 |
InChI Key | RDYMFSUJUZBWLH-VQDHCCOSSA-N |
SMILES | ClC1=C(Cl)[C@@]2(Cl)[C@H]3COS(=O)OC[C@H]3C1(Cl)C2(Cl)Cl |
IUPAC Name | (1S,2R,8S)-1,9,10,11,12,12-hexachloro-4,6-dioxa-5λ⁴-thiatricyclo[7.2.1.0²,⁸]dodec-10-en-5-one |
Show More |
Safety and Handling
Product Identifier
- Standard solution Endosulfan-beta 100ug/ml in acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P330-Rinse mouth.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'