Learn More
Atrazine in Acetonitrile 100μg/mL, Fisher Chemical™
BIOPHARMA
Product Code. 12979543 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12979543 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12979543 Supplier Fisher Chemical Supplier No. P804990
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule
CAS 1912-24-9
Physical Form Liquid
Molecular Formula C8H14ClN5
Concentration 100 μg/mL
MDL Number MFCD00041810
UN Number 1648
InChI Key MXWJVTOOROXGIU-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CCNC1=NC(Cl)=NC(NC(C)C)=N1
IUPAC Name 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 215.69
PubChem CID 2256
Quantity 1 mL
ChEBI CHEBI:15930
Chemical Name or Material Atrazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Atrazine,2-chloro-4-ethylamine-6-isopropylamine-1,3,5-triazine 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .