Learn More
Meta-Topolin, TRC
Specifications
Specifications
| Chemical Name or Material | Meta-Topolin |
| CAS | 75737-38-1 |
| Molecular Formula | C12H11N5O |
| Quantity | 5 mg |
| Synonym | 6-m-Hydroxybenzylaminopurine, 3-[(9H-Purin-6-ylamino)methyl]phenol, 6-(3-Hydroxybenzylamino)purine, m-Topolin |
| InChI Formula | InChI=1S/C12H11N5O/c18-9-3-1-2-8(4-9)5-13-11-10-12(15-6-14-10)17-7-16-11/h1-4,6-7,18H,5H2,(H2,13,14,15,16,17) |
| SMILES | Oc1cccc(CNc2ncnc3nc[nH]c23)c1 |
| IUPAC Name | 3-[(7H-purin-6-ylamino)methyl]phenol |
| Molecular Weight (g/mol) | 241.25 |
| Formula Weight | 241.0964 |