Learn More
Mestranol, MedChemExpress
Product Code. 16488276
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16488276 Supplier Medchem Express Supplier No. HYB0390
 Restricted Item
Estimated Shipment: 18-01-2027
MedChemExpress Mestranol is an inactive prodrug and becomes biologically active on conversion to ethinyl estradiol (EE). Mestranol acts as an estrogen receptor agonist. Mestranol combines with a progestin in vivo and can be used for the research of menopausal hormone or menstrual disorders.
Mestranol is an inactive prodrug and becomes biologically active on conversion to ethinyl estradiol (EE). Mestranol acts as an estrogen receptor agonist. Mestranol combines with a progestin in vivo and can be used for the research of menopausal hormone or menstrual disorders.
Specifications
Chemical Name or Material Mestranol
CAS 72-33-3
Color White
Purity Grade Notes Research
Molecular Formula C21H26O2
Quantity 100 mg
Solubility Information DMSO : 33.33 mg/mL (107.37 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.16 mM; Need ultrasonic)
SMILES C[C@@]1([C@]2(O)C#C)[C@](CC2)([H])[C@@](CCC3=C4C=CC(OC)=C3)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 310.43
Formula Weight 310.43
Percent Purity 99.88%
Grade Research
Physical Form Solid