Learn More
Mesitylenesulfonic acid dihydrate, 98+%
Product Code. 11466996
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11466996 100 g 100g
11456996 25 g 25g
2 options
Product Code. 11466996
Brand: Thermo Scientific Alfa Aesar B25321.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 835617-36-2 | C9H16O5S | 236.282 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 835617-36-2
Molecular Formula C9H16O5S
Molecular Weight (g/mol) 236.282
MDL Number MFCD00149535
InChI Key LIKWMDGAWXCECN-UHFFFAOYSA-N
Synonym mesitylenesulfonic acid dihydrate, 2-mesitylenesulfonic acid dihydrate, 2,4,6-trimethylbenzenesulfonic acid dihydrate, benzenesulfonic acid, 2,4,6-trimethyl-, dihydrate, mesitylenesulfonicaciddihydrate, mesitylenesulfonic acid dihydrate, 98+%, benzenesulfonic acid,2,4,6-trimethyl-, hydrate 1:2, 2,4,6-trimethylbenzenesulfonic acid, oxamethane, oxamethane
PubChem CID 15620344
IUPAC Name 2,4,6-trimethylbenzenesulfonic acid;dihydrate
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)O)C.O.O
Specifications
Melting Point 74°C to 77°C
Quantity 100 g
UN Number UN2585
Beilstein 1961501
Sensitivity Hygroscopic
Formula Weight 236.29 (200.26 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Mesitylenesulfonic acid dihydrate