Learn More
Mesembrine, TRC
Product Code. 17963365
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963365 10 mg 10mg
17914155 1 mg 1mg
17962565 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17963365 Supplier Toronto Research Chemicals Supplier No. TRCM33895010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24880-43-1
Molecular Formula C17H23NO3
Molecular Weight (g/mol) 289.37
Synonym (3aS,7aS)-3a-(3,4-Dimethoxyphenyl)octahydro-1-methyl-6H-indol-6-one, 4α,9α-Mesembrine, (3aS-cis)-3a-(3,4-Dimethoxyphenyl)octahydro-1-
SMILES COc1ccc(cc1OC)[C@]23CCN(C)[C@H]2CC(=O)CC3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H23NO3/c1-18-9-8-17(7-6-13(19)11-16(17)18)12-4-5-14(20-2)15(10-12)21-3/h4-5,10,16H,6-9,11H2,1-3H3
Formula Weight 289.1678
Percent Purity >95%
Chemical Name or Material Mesembrine
Grade HPLC