missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Bromide, Supelco™

Product Code. 15894430
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894430 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15894430 Supplier Supelco Supplier No. 1044210050
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-47-1

Specifications

Boiling Point 322°C/1013 hPa (sublimed)
Melting Point 237°C
Quantity 50 g
MDL Number MFCD00011040
Synonym Mercury(II) bromide, Mercury(II) bromide
Molecular Weight (g/mol) 360.4
Grade Research
Density 6.1 g/cm3 at 25°C
Chemical Name or Material Mercury(II) Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.