Learn More
β-Mercaptoethylamine hydrochloride, ∽98%, MP Biomedicals™
Product Code. 11388569 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388569 25 g 25g
11378569 5 g 5g
11398569 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11388569 Supplier MP Biomedicals Supplier No. 0210099525
  • ONLINE EXCLUSIVE PRICE
    110.00 EUR
    103.95 EUR / 25g
    Save 6.05 EUR
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Chemical Identifiers
CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
SMILES [H+].[Cl-].NCCS
Specifications
Color White
Assay Percent Range ∽ 98%
Quantity 25 g
IUPAC Name hydrogen 2-aminoethane-1-thiol chloride
Formula Weight 113.6
Physical Form Powder
Chemical Name or Material β-Mercaptoethylamine hydrochloride