Learn More
Mepivacaine, TRC
Product Code. 30135927
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135927 1 g 1g
17972925 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30135927 Supplier Toronto Research Chemicals Supplier No. TRCM2250781G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 96-88-8
Molecular Formula C15 H22 N2 O
Molecular Weight (g/mol) 246.35
Synonym 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-1-methyl-, 2',6'-Pipecoloxylidide, 1-methyl- (6CI,8CI), N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide, (±)-Mepivacaine, 1-Methyl-2',6'-pipecoloxylidide, APF 135, Carbocain, Carbocaine, Carbocaine-V, DL-Mepivacaine, MepiSV, Mepicaine, Mepivacaine, Scandicain, Scandicaine, Tevacaine
IUPAC Name N-(2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide
SMILES CN1CCCCC1C(=O)Nc2c(C)cccc2C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C15H22N2O/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18)
Formula Weight 246.1732
Percent Purity >95%
Chemical Name or Material Mepivacaine
Grade HPLC