Learn More
Mepivacaine, Lipomed
Product Code. 30461907
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461907 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461907 Supplier Lipomed Supplier No. LPMMPV1872FB100
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Mepivacaine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mepivacaine
CAS 96-88-8
Analyte or Component Names Mepivacaine
Shipping Condition Room Temperature
Molecular Formula C15 H22 N2 O
Synonym 2-Piperidinecarboxamide, N-(2,6-dimethylphenyl)-1-methyl-, 2',6'-Pipecoloxylidide, 1-methyl- (6 CI,8 CI), N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide, (±)-Mepivacaine, 1-Methyl-2',6'-pipecoloxylidide, APF 135, Carbocain, Carbocaine, Carbocaine-V, DL-Mepivacaine, MepiSV, Mepicaine, Mepivacaine, Scandicain, Scandicaine, Tevacaine
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN1CCCCC1C(=O)Nc2c(C)cccc2C
IUPAC Name N-(2,6-dimethylphenyl)-1-methyl-piperidine-2-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 246.35
Quantity 100 mg
Formula Weight 246.1732 g/mol