Learn More
(-)-Menthol, Sigma-Aldrich™
Specifications
Specifications
| CAS | 2216-51-5 |
| Melting Point | 41°C to 44°C |
| Density | 0.890 g/cm3 at 20°C |
| Boiling Point | 216°C/1013 hPa |
| MDL Number | MFCD00062979 |
| Quantity | 100 g |
| Synonym | (-)-Menthol, 2-Isopropyl-5-methylcyclohexanol, (-)-Menthol |
| Molecular Weight (g/mol) | 156.27 |
| Grade | Research |