Learn More
Fisherbrand™ Traceable™ Memory-Loc™ Data Logging Thermohygrometers with Calibration
Product Code. 16641323 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Data Memory:
819,150 readings
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Data Memory Unit Size
16641323 819,150 readings Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16641323
  • ONLINE EXCLUSIVE PRICE
    344.00 EUR
    263.68 EUR / Pack of 1
    Save 80.32 EUR
    23% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
Safely secure collected information. Data cannot be overwritten or modified.
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Store up to 30 unique alarm events
  • Smart-Alarm™ with visual/audible alarm continues to alert even if the unit returns to nonalarm conditions
  • Adjustable sampling rate—from 1 min to 12 hrs in 1 min steps
Specifications
Type Thermohygrometer with Barometer and Calibration
Data Memory 819,150 readings
Calibration NIST
Channels 1
Accuracy (Relative Humidity) ±3% RH between 5 to 75%, otherwise ±5% RH
Accuracy (Temperature) ±0.4°C
Humidity Range 0 to 95%, non-condensing
Resolution (Relative Humidity) 0.1% RH
Temperature Resolution 0.1°C
Temperature Range (Metric) 0°C to 65°C